Learn More
7-Aminoheptanoic Acid Ethyl Ester Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 7-Aminoheptanoic Acid Ethyl Ester Hydrochloride |
| CAS | 29840-65-1 |
| Molecular Formula | C9 H19 N O2 . Cl H |
| Quantity | 5 g |
| Synonym | Ethyl 7-Aminoheptanoate Hydrochloride |
| InChI Formula | InChI=1S/C9H19NO2.ClH/c1-2-12-9(11)7-5-3-4-6-8-10;/h2-8,10H2,1H3;1H |
| SMILES | Cl.CCOC(=O)CCCCCCN |
| IUPAC Name | ethyl 7-aminoheptanoate;hydrochloride |
| Molecular Weight (g/mol) | 209.71 |
| Formula Weight | 209.1183 |