missing translation for 'onlineSavingsMsg'
Learn More

(6E)-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3-hydroxy-5-oxo-6-heptenoic Acid 1,1-Dimethylethyl Ester, TRC

Product Code. 30403342
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403342 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403342 Supplier Toronto Research Chemicals Supplier No. TRCF62172310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (6E)-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3-hydroxy-5-oxo-6-heptenoic Acid 1,1-Dimethylethyl Ester
CAS 1379516-02-5
Molecular Formula C19 H29 N3 O6
Quantity 10 mg
Synonym (E)-t-Butyl 7-(3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl)-3-hydroxy-5-oxohept-6-enoate
InChI Formula InChI=1S/C19H29N3O6/c23-16(13-20-7-8-21-19(26)22-9-11-27-12-10-22)14-28-17-4-1-15(2-5-17)3-6-18(24)25/h1-2,4-5,16,20,23H,3,6-14H2,(H,21,26)(H,24,25)/t16-/m0/s1
SMILES O[C@@H](CNCCNC(=O)N1CCOCC1)COc2ccc(CCC(=O)O)cc2
IUPAC Name 3-[4-[(2S)-2-hydroxy-3-[2-(morpholine-4-carbonylamino)ethylamino]propoxy]phenyl]propanoic acid
Molecular Weight (g/mol) 395.45
Formula Weight 395.206
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.