Learn More
6Alpha-Methylprednisolone 21-Hemisuccinate, TRC
Product Code. 30136059
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
100 mg
Unit Size:
100mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136059 25 mg 25mg
30135647 100 mg 100mg
30041219 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136059 Supplier Toronto Research Chemicals Supplier No. TRCM33119525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2921-57-5
Molecular Formula C26 H34 O8
Molecular Weight (g/mol) 474.54
Synonym Methylprednisolone hydrogen succinate, 4-[(11β,17-Dihydroxy-6α-methyl-3,20-dioxopregna-1,4-dien-21-yl)oxy]-4-oxobutanoic acid, (6α,11β)-21-(3-carboxy-1-oxopropoxy)-11,17-dihydroxy-6-methyl-pregna-1,4-diene-3,20-dione, Pregna-1,4-diene-3,20-dione, 11β,17,21-trihydroxy-6α-methyl-, 21-(H succinate) (7CI), Pregna-1,4-diene-3,20-dione, 11β,17,21-trihydroxy-6α-methyl-, 21-(hydrogen succinate) (8CI), Pregna-1,4-diene-3,20-dione, 11β,17,21-trihydroxy-6α-methyl-, 21-succinate (6CI), 6-Methylprednisolone hemisuccinate, 6α-Methylprednisolone 21-succinate, 6α-Methylprednisolone hemisuccinate, Methylprednisolone 21-hemisuccinate, Methylprednisolone hemisuccinate, Methylprednisolone succinate
IUPAC Name 4-[2-[(6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-6,10,13-trimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-4-oxobutanoic acid
SMILES C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)COC(=O)CCC(=O)O)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]4(C)C=CC(=O)C=C14
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C26H34O8/c1-14-10-16-17-7-9-26(33,20(29)13-34-22(32)5-4-21(30)31)25(17,3)12-19(28)23(16)24(2)8-6-15(27)11-18(14)24/h6,8,11,14,16-17,19,23,28,33H,4-5,7,9-10,12-13H2,1-3H3,(H,30,31)/t14-,16-,17-,19-,23+,24-,25-,26-/m0/s1
Formula Weight 474.2254
Chemical Name or Material Methylprednisolone Hydrogen Succinate