Learn More
6Alpha-Methyl-9Alpha-fluoro-11Beta,17Alpha-dihydroxyprogesterone, TRC
Specifications
Specifications
| Chemical Name or Material | 6alpha-Methyl-9alpha-fluoro-11beta,17alpha-dihydroxyprogesterone |
| CAS | 378-36-9 |
| Molecular Formula | C22H31FO4 |
| Quantity | 5 mg |
| Synonym | 6α-Methyl-9α-fluoro-21-desoxyhydrocortisone, U 8390 |
| InChI Formula | InChI=1S/C19H14O/c1-20-15-8-11-17-14(12-15)7-10-18-16-5-3-2-4-13(16)6-9-19(17)18/h2-12H,1H3 |
| SMILES | COC1=CC=C(C(C=C2)=C1)C3=C2C4=CC=CC=C4C=C3 |
| Molecular Weight (g/mol) | 378.48 |