Learn More
Masterflex™ 65mm Correlated PTFE Flowmeter
Product Code. 11735259 Shop All Masterflex Products
Change view
Click to view available options
Pressure Rating:
100 psig
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pressure Rating Unit Size
11735259 100 psig Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11735259 Supplier Masterflex™ Supplier No. 3204457
  • ONLINE EXCLUSIVE PRICE
    587.00€
    454.37€ / Pack of 1
    Save 132.63€
    22% Off
Estimated Shipment: 02-12-2026
Specifications
Type Variable Area Flowmeter
Accuracy ±2% (Full Scale)
Float Material PTFE, Glass
Pressure Rating 100 psig
Connections 1/8 in. NPT (F)
Flow Rate Range 122 mL/min. CO2, 94 mL/min. O2, 113 mL/min. N2, 86 mL/min. Ar, 284 mL/min. H2
For Use With (Application) For high-purity applications with flexibility for a broad array of gases
Repeatability ±0.25%
Scale 65 mm
Temperature (Metric) 65°C