Learn More
Jenway™ 6320 Benchtop Visible Spectrophotometer
Click to view available options
Spectral Bandwidth:
8 nm, 6 nm over the UV range
missing translation for 'unitSize'
Each
Description
- Recalls previous settings at power up
- Analog and RS-232 outputs let you interface with printers and PCs
- Dual-line LCD shows error messages, prompts, mode indication, and concentration units
Specifications
Specifications
| Light Source | Xenon Flash |
| Spectral Bandwidth | 8 nm, 6 nm over the UV range |
| For Use With (Application) | Education, quality control, environmental and clinical analysis |
| Includes | Mains lead, Pk. 100 disposable cuvettes, 10 x 10 mm cell holder, interface cable and PC application software on CD-ROM |
| Optical Design | Independently housed and isolated with lenses |