Learn More
3M™ 6000 Series Half Face Mask Respirator
Description
- Reusable half masks are economical, low-maintenance, simple-to-handle, and extremely lightweight
- Used with 3M™ Supplied Air Systems
- Bayonet Connection System allows connection to a broad range of twin lightweight filters to protect against gases, vapors, and particulates
- Twin filter design provides lower breathing resistance
- Probed reusable respirator assembly 6000 Series used for quantitative fit testing (17-987-3A & -3B)
Compliance
NIOSH approved under 42 CFR 84 standard for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters; in Canada, CSA standard Z94.4 and applicable local government requirements must be met for approval.
Specifications
Specifications
| Color | Gray |
| Material | Thermoplastic Elastomer |
| Product Type | Half-Mask Respirator |
| For Use With (Application) | Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding |
| Model | K6200 |
| Suspension Type | 4-point |