Learn More
Greiner Bio-One 60 Well HLA Terasaki Plates
GREENER_CHOICE
Product Code. 10318801
Click to view available options
Quantity:
270/Cs.
Unit Size:
Pack of 270
Product Code. 10318801
Brand: Greiner Bio-One 653180
Only null left
Add to basket
Add to basket
Suitable for all applications for serological determination of HLA antigens
  • Suitable for all applications for serological determination of HLA antigens
  • Manufactured from crystal clear polystyrene
  • Stackable
  • Improved cell adhesion through physical surface treatment
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Autoclavable Not Autoclavable
Bottom Shape V
Capacity (Metric) 10 L (Well)
Color Clear
Material Polystyrene
Shape Rectangular
Sterility Non-sterile
Surface Treatment Tissue Culture
Type Microtest Plate
Well Count 60
For Use With (Application) HLA testing
Includes Lid
No. per Case 10 pcs/bag
No. per Pack 10
Packaging Inner bags of 10
Quantity 270/Cs.
Temperature Range (Metric) -20°C to +60°C
Dimensions (L x W x H) 83 mm x 58 mm x 11 mm
Length (Metric) 83.3 mm
Width (Metric) 58 mm
Height (Metric) 11 mm