Learn More
BRAND™ 6-well Insert 2in1, PET-Membrane
Product Code. 16194967 Shop All Brand Products
Click to view available options
Pore Size:
0.4 μm
8 μm
No. per Pack:
24 x 1
4 x 6
Unit Size:
Pack of 24
Product Code. 16194967
Brand: BRAND™ 782757
Only null left
Add to basket
Add to basket
The cellGrade™ plus surface of the membranes ensures excellent cell attachment and growth. The highly transparent membranes give you good visibilty of your cell cultures, and the high pore density contributes to good permeability.
  • Use Inserts in a hanging or standing position
  • For 6-well plates or culture dishes
  • Achieve uniform and controlled cell growth due to surface treatment, without coatings
TRUSTED_SUSTAINABILITY
Specifications
Description 6-well Insert 2in1, PET-Membrane 8 μm, Multipack
Material PET
Pore Size 8 μm
No. per Pack 4 x 6
Sterility Sterile
No. of Wells 6