Learn More
6-Phenylphenathridine-3,8-diamine, TRC
Product Code. 30414545
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414545 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30414545 Supplier Toronto Research Chemicals Supplier No. TRCP3358601G
 Restricted Item
Estimated Shipment: 04-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-Phenylphenathridine-3,8-diamine
CAS 52009-64-0
Molecular Formula C19 H15 N3
Quantity 1 g
InChI Formula InChI=1S/C19H15N3/c20-13-6-8-15-16-9-7-14(21)11-18(16)22-19(17(15)10-13)12-4-2-1-3-5-12/h1-11H,20-21H2
SMILES Nc1ccc2c(c1)nc(c3ccccc3)c4cc(N)ccc24
IUPAC Name 6-phenylphenanthridine-3,8-diamine
Molecular Weight (g/mol) 285.34
Formula Weight 285.1266