Learn More
6-Pentyltetrahydro-2H-pyran-2-one, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 705-86-2 |
| Molecular Formula | C10H18O2 |
| MDL Number | MFCD00006649 |
| Quantity | 100 g |
| SMILES | O=C1CCCC(CCCCC)O1 |
| Molecular Weight (g/mol) | 170.25 |
| Percent Purity | 97% |
| Chemical Name or Material | 6-Pentyltetrahydro-2H-pyran-2-one |