Learn More
6-Methyl-hepten-2-one, TRC
Product Code. 17913295
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17913295 25 g 25g
17953945 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17913295 Supplier Toronto Research Chemicals Supplier No. TRCM31190025G
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 110-93-0
Molecular Formula C8 H14 O
Molecular Weight (g/mol) 126.1962
Synonym Isoprenylacetone, 5-Hepten-2-one, 6-methyl-, 6-Methyl-5-hepten-2-one, 2-Methyl-2-hepten-6-one, 2-Methyl-2-heptene-6-ketone, 2-Methyl-2-heptene-6-one, 2-Methyl-6-oxo-2-heptene, 2-Oxo-6-methylhept-5-ene, 6-Methyl-5-hepten-2-ketone, 6-Methyl-5-heptene-2-one, 6-Methyl-δ5-hepten-2-one, Isoprenylacetone, Methylheptenone, NSC 15294, NSC 66569, Prenylacetone, Sulcatone, 2-Methyl-2-hepten-6-one, 2-Methyl-2-heptene-6-ketone, 2-Methyl-2-heptene-6-one, 2-Methyl-6-oxo-2-heptene, 2-Oxo-6-methylhept-5-ene, 6-Methyl-5-hepten-2-ketone, 6-Methyl-5-hepten-2-one, 6-Methyl-5-heptene-2-one, 6-Methyl-δ5-hepten-2-one, Isoprenylacetone, Methylheptenone, NSC 15294, NSC 66569, Prenylacetone, Sulcatone
IUPAC Name 6-methylhept-5-en-2-one
SMILES CC(=CCCC(=O)C)C
Specifications
Quantity 25 g
InChI Formula InChI=1S/C8H14O/c1-7(2)5-4-6-8(3)9/h5H,4,6H2,1-3H3
Formula Weight 126.1045
Chemical Name or Material 6-Methyl-hepten-2-one