Learn More
6-Methyl-2-phenyl-4H-chromen-4-one, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 29976-75-8 |
| Molecular Formula | C16H12O2 |
| MDL Number | MFCD00017461 |
| Quantity | 1 g |
| SMILES | O=C1C=C(C2=CC=CC=C2)OC3=C1C=C(C)C=C3 |
| Molecular Weight (g/mol) | 236.27 |
| Percent Purity | 99% |
| Chemical Name or Material | 6-Methyl-2-phenyl-4H-chromen-4-one |