Learn More
6-Mercaptopurine-9-Beta-D-ribofuranoside 2',3',5'-Triacetate, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Mercaptopurine-9-beta-D-ribofuranoside 2',3',5'-Triacetate |
| CAS | 3021-21-4 |
| Molecular Formula | C16H18N4O7S |
| Quantity | 10 g |
| Synonym | 9-beta-D-ribofuranosyl-9H-purine-6-thiol 2’,3’,5’-Triacetate, 6-Thioinosine 2',3',5'-triacetate, NSC 66385, (2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(6-mercapto-9H-purin-9-yl)tetrahydrofuran-3,4-diyl Diacetate |
| InChI Formula | InChI=1S/C16H18N4O7S/c1-7(21)24-4-10-12(25-8(2)22)13(26-9(3)23)16(27-10)20-6-19-11-14(20)17-5-18-15(11)28/h5-6,10,12-13,16H,4H2,1-3H3,(H,17,18,28)/t10-,12-,13-,16-/m1/s1 |
| SMILES | CC(=O)OC[C@H]1O[C@H]([C@H](OC(=O)C)[C@@H]1OC(=O)C)n2cnc3c(S)ncnc23 |
| IUPAC Name | [(2R,3R,4R,5R)-3,4-diacetyloxy-5-(6-sulfanylpurin-9-yl)oxolan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 410.4 |
| Formula Weight | 410.0896 |