missing translation for 'onlineSavingsMsg'
Learn More

6-Keto Ethynyl Estradiol, TRC

Product Code. 30040962
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040962 10 mg 10mg
30006078 50 mg 50mg
17979959 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040962 Supplier Toronto Research Chemicals Supplier No. TRCK18877010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38002-18-5
Molecular Formula C20 H22 O3
Molecular Weight (g/mol) 310.39
Synonym 3,17-Dihydroxy-19-nor-17alpha-pregna-1,3,5(10)-trien-20-yn-6-one, Ethinylestradiol Imp. G (EP), 6-Oxoethinylestradiol, 6-Ketoethinylestradiol
IUPAC Name (8R,9S,13S,14S,17R)-17-ethynyl-3,17-dihydroxy-13-methyl-7,8,9,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-6-one
SMILES C[C@]12CC[C@H]3[C@@H](CC(=O)c4cc(O)ccc34)[C@@H]1CC[C@@]2(O)C#C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H22O3/c1-3-20(23)9-7-17-15-11-18(22)16-10-12(21)4-5-13(16)14(15)6-8-19(17,20)2/h1,4-5,10,14-15,17,21,23H,6-9,11H2,2H3/t14-,15-,17+,19+,20+/m1/s1
Formula Weight 310.1569
Percent Purity >95%
Chemical Name or Material 3,17-Dihydroxy-19-nor-17alpha-pregna-1,3,5(10)-trien-20-yn-6-one (6-Oxoethinylestradiol; 6-Ketoethinylestradiol)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.