Learn More
6-Heptyn-1-ol, 95%
Product Code. 15468068
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468068 5 g 5g
15458068 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15468068 Supplier Thermo Scientific Alfa Aesar Supplier No. H61509.06
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
CAS: 63478-76-2 | C7H12O | 112.17 g/mol
6-Heptyn-1-ol is used as a reagent in in the preparation of many cyclic compounds. It is also used as a Intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Heptyn-1-ol is used as a reagent in in the preparation of many cyclic compounds. It is also used as a Intermediate in organic synthesis.
Solubility
Soluble in chloroform, dichloromethane and methanol. Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 63478-76-2
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00049198
InChI Key BVRCLEXKQNWTDK-UHFFFAOYSA-N
Synonym 6-heptyn-1-ol, 7-hydroxyhept-1-yne, pubchem13080, ho ch2 5c=ch, ksc352m8p
PubChem CID 11007849
IUPAC Name hept-6-yn-1-ol
SMILES OCCCCCC#C
Specifications
Linear Formula HC≡C(CH2)5OH
Quantity 5 g
Solubility Information Soluble in chloroform,dichloromethane and methanol. Slightly soluble in water.
Formula Weight 112.17
Percent Purity 95%
Chemical Name or Material 6-Heptyn-1-ol