Learn More
6-Fluorothiochroman-4-one, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 21243-18-5 |
| Molecular Formula | C9H7FOS |
| MDL Number | MFCD00078488 |
| Quantity | 25 g |
| SMILES | O=C1CCSC2=C1C=C(F)C=C2 |
| Molecular Weight (g/mol) | 182.21 |
| Percent Purity | 97% |
| Chemical Name or Material | 6-Fluorothiochroman-4-one |