Learn More
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride, 98%, BLD Pharm™
Product Code. 31450393
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31450393 25 g
31449425 100 g
31442762 5 g
31439852 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31450393 Supplier BLD Pharm Supplier No. BD1391225g
 Restricted Item
Estimated Shipment: 25-11-2026
Specifications
CAS 84163-13-3
Molecular Formula C12H14ClFN2O
MDL Number MFCD06658529
Quantity 25 g
SMILES FC1=CC2=C(C=C1)C(C3CCNCC3)=NO2.[H]Cl
Molecular Weight (g/mol) 256.70
Percent Purity 98%
Chemical Name or Material 6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride