missing translation for 'onlineSavingsMsg'
Learn More

6-Dehydrocortisol Acetate, TRC

Product Code. 30367676
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367676 25 mg 25mg
30359694 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30367676 Supplier Toronto Research Chemicals Supplier No. TRCD22922525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21940-45-4
Molecular Formula C23 H30 O6
Molecular Weight (g/mol) 402.48
Synonym Pregna-4,6-diene-3,20-dione, 21-(acetyloxy)-11,17-dihydroxy-, (11β)- (9CI), Pregna-4,6-diene-3,20-dione, 11β,17,21-trihydroxy-, 21-acetate (6CI,7CI,8CI), δ6-Cortisol 21-acetate, 11β,17,21-Trihydroxypregna-4,6-diene-3,20-dione 21-Acetate
IUPAC Name [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3C=CC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C23H30O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h4-5,10,16-18,20,26,28H,6-9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1
Formula Weight 402.2042
Chemical Name or Material 11beta,17,21-Trihydroxypregna-4,6-diene-3,20-dione 21-Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.