Learn More
6-chloroimidazo[1,2-b]pyridazine-2-carboxylic acid, 90%, Thermo Scientific™
Product Code. p-3075844
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10279082 250 mg 250mg
10520113 1 g 1g
10332115 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10279082 Supplier Thermo Scientific Maybridge Supplier No. MO00885CB
Chemical Identifiers
CAS 14714-24-0
Molecular Formula C7H4ClN3O2
Molecular Weight (g/mol) 197.578
MDL Number MFCD04974055
InChI Key HTUZJYQXKFDVLT-UHFFFAOYSA-N
Synonym 6-chloroimidazo 1,2-b pyridazine-2-carboxylic acid, 2-carboxy-6-chloroimidazo 1,2-b pyridazine, 6-chloro-imidazo 1,2-b pyridazine-2-carboxylic acid, pubchem23037, 6-chloroimidazo 1,2-b pyridazine-2-carboxylicacid, 6-chloro-2-imidazo 1,2-b pyridazinecarboxylic acid, 6-chloranylimidazo 1,2-b pyridazine-2-carboxylic acid, imidazo 1,2-b pyridazine-2-carboxylicacid, 6-chloro, 6-chloro-4-hydroimidazo 1,2-e pyridazine-2-carboxylic acid, imidazo 1,2-b pyridazine-2-carboxylic acid, 6-chloro
PubChem CID 2794637
IUPAC Name 6-chloroimidazo[1,2-b]pyridazine-2-carboxylic acid
SMILES C1=CC(=NN2C1=NC(=C2)C(=O)O)Cl
Specifications
CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 197.58
Percent Purity 90%
Chemical Name or Material 6-Chloroimidazo[1, 2-b]pyridazine-2-carboxylic acid