Learn More
6-Chloro-6-deoxy-alpha-D-glucopyranose, TRC
Product Code. 30360700
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30360700 50 mg 50mg
30359953 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30360700 Supplier Toronto Research Chemicals Supplier No. TRCC36776050MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28528-86-1
Molecular Formula C6H11ClO5
Molecular Weight (g/mol) 198.6
Synonym (2S,3R,4S,5S,6S)-6-(Chloromethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol
IUPAC Name (2S,3R,4S,5S,6S)-6-(chloromethyl)oxane-2,3,4,5-tetrol
SMILES O[C@H]1O[C@H](CCl)[C@@H](O)[C@H](O)[C@H]1O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C6H11ClO5/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-6,8-11H,1H2/t2-,3-,4+,5-,6+/m1/s1
Formula Weight 198.0295
Chemical Name or Material 6-Chloro-6-deoxy-alpha-D-glucopyranose