missing translation for 'onlineSavingsMsg'
Learn More

6-Bromopyridine-3-boronic acid, 95%

Product Code. 11384516
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11384516

Brand: Thermo Scientific Alfa Aesar L20085.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 223463-14-7 | C5H5BBrNO2 | 201.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 223463-14-7
Molecular Formula C5H5BBrNO2
Molecular Weight (g/mol) 201.81
MDL Number MFCD03411558
InChI Key BCYWDUVHAPHGIP-UHFFFAOYSA-N
Synonym 2-bromopyridine-5-boronic acid, 6-bromopyridin-3-yl boronic acid, 6-bromopyridine-3-boronic acid, 2-bromo-5-pyridineboronic acid, 6-bromopyridin-3-lboronic acid, 2-bromo-5-pyridylboronic acid, 2-bromopyridyl-5-boronic acid, 6-bromo-3-pyridylboronic acid, 6-bromopyridin-3-yl-3-boronic acid, 6-bromo-3-pyridinylboronic acid
PubChem CID 2762706
IUPAC Name (6-bromopyridin-3-yl)boronic acid
SMILES OB(O)C1=CN=C(Br)C=C1

Specifications

Melting Point ∽165°C (decomposition)
Quantity 1 g
Beilstein 9119918
Formula Weight 201.82
Percent Purity 95%
Chemical Name or Material 6-Bromopyridine-3-boronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.