Learn More
6-Bromopicolinamide, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 25194-52-9 |
| Molecular Formula | C6H5BrN2O |
| MDL Number | MFCD00234154 |
| Quantity | 5 g |
| SMILES | O=C(N)C1=NC(Br)=CC=C1 |
| Molecular Weight (g/mol) | 201.02 |
| Percent Purity | 98% |
| Chemical Name or Material | 6-Bromopicolinamide |