Learn More
6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid, 97%, Thermo Scientific™
Product Code. 15522303 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15522303 1 g 1g
15532303 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15522303 Supplier Thermo Scientific Acros Supplier No. 449910010
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Chemical Identifiers
CAS 944896-42-8
Molecular Formula C8H5BrN2O2
Molecular Weight (g/mol) 241.04
MDL Number MFCD10698063
InChI Key SDKJLYHPCBMDAD-UHFFFAOYSA-N
Synonym 6-bromoimidazo 1,2-a pyridine-3-carboxylic acid, 6-bromoimidazo 1,2-a pyridine-3-carboxylicacid, 6-bromoh-imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo-1h-imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo-3-carboxyimidazo 1,2-a pyridine, 6-bromo-imidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo
PubChem CID 22031912
IUPAC Name 6-bromoimidazo[1,2-a]pyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=C2C=CC(Br)=CN12
Specifications
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 241.04
Percent Purity 97%
Chemical Name or Material 6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid
compliance-icons
Product Identifier
  • 6-Bromoimidazo¢1,2-a!pyridine-3-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.