missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-methoxyquinoline, 96%

Product Code. p-7035991
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15430568 250 mg 250mg
15410568 1 g 1g
15420568 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15430568 Supplier Thermo Scientific Alfa Aesar Supplier No. H54226.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 99455-05-7 | C10H8BrNO | 238.084 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99455-05-7
Molecular Formula C10H8BrNO
Molecular Weight (g/mol) 238.084
MDL Number MFCD11847805
InChI Key KBTKKEMYFUMFSJ-UHFFFAOYSA-N
Synonym 2-methoxy-6-bromoquinoline, 6-bromo-2-methoxy-quinoline, quinoline, 6-bromo-2-methoxy, 6-bromanyl-2-methoxy-quinoline
PubChem CID 10657538
IUPAC Name 6-bromo-2-methoxyquinoline
SMILES COC1=NC2=C(C=C1)C=C(C=C2)Br

Specifications

Melting Point 87°C to 89°C
Quantity 250 mg
Formula Weight 238.08
Percent Purity 96%
Chemical Name or Material 6-Bromo-2-methoxyquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.