Learn More
6-Bromo-2-(4-bromophenyl)indole, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Bromo-2-(4-bromophenyl)indole |
| CAS | 28718-96-9 |
| Molecular Formula | C14H9Br2N |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C14H9Br2N/c15-11-4-1-9(2-5-11)13-7-10-3-6-12(16)8-14(10)17-13/h1-8,17H |
| SMILES | Brc1ccc(cc1)c2cc3ccc(Br)cc3[nH]2 |
| IUPAC Name | 6-bromo-2-(4-bromophenyl)-1H-indole |
| Molecular Weight (g/mol) | 351.04 |
| Formula Weight | 348.9102 |