missing translation for 'onlineSavingsMsg'
Learn More

6-Benzyloxy-1-cyclopropyl-1,4-dihydro-7-fluoro-4-oxo-3-quinolinecarboxylic Acid Benzyl Ester, TRC

Product Code. 30335880
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335880 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335880 Supplier Toronto Research Chemicals Supplier No. TRCB28798525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Benzyloxy-1-cyclopropyl-1,4-dihydro-7-fluoro-4-oxo-3-quinolinecarboxylic Acid Benzyl Ester
CAS 1391053-23-8
Molecular Formula C27H22FNO4
Quantity 25 mg
InChI Formula InChI=1S/C27H22FNO4/c28-23-14-24-21(13-25(23)32-16-18-7-3-1-4-8-18)26(30)22(15-29(24)20-11-12-20)27(31)33-17-19-9-5-2-6-10-19/h1-10,13-15,20H,11-12,16-17H2
SMILES Fc1cc2N(C=C(C(=O)OCc3ccccc3)C(=O)c2cc1OCc4ccccc4)C5CC5
IUPAC Name benzyl 1-cyclopropyl-7-fluoro-4-oxo-6-phenylmethoxyquinoline-3-carboxylate
Molecular Weight (g/mol) 443.47
Formula Weight 443.1533
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.