missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 6-Aminopenicillanic acid, 98%
Click to view available options
Quantity:
10g
50g
250g
Unit Size:
10g
250g
50g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
6-Aminopenicillanic acid is the basic unit in penicillin’s, which is obtained from the fermentation brew of the Penicillium mold. It acts as a starting material for the preparation of many semisynthetic penicillins. Further, it serves as apharmaceutical intermediate. In addition to this, it is used in antibiotics research and experimental application.
Solubility
Soluble in water and hydrochoric acid.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
6-Aminopenicillanic acid is the basic unit in penicillin’s, which is obtained from the fermentation brew of the Penicillium mold. It acts as a starting material for the preparation of many semisynthetic penicillins. Further, it serves as apharmaceutical intermediate. In addition to this, it is used in antibiotics research and experimental application.
Solubility
Soluble in water and hydrochoric acid.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Purity | 98% |
| CAS | 551-16-6 |
| Content And Storage | Keep cold |
| Molecular Weight (g/mol) | 216.26 |
| Product Type | 6-Aminopenicillanic acid |
| Quantity | 50g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction