Learn More
6-Aminocaproic Acid Dimer, TRC
Product Code. 30010183
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010183 25 mg 25mg
30009495 250 mg 250mg
30005275 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010183 Supplier Toronto Research Chemicals Supplier No. TRCA60301825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2014-58-6
Molecular Formula C12H24N2O3
Molecular Weight (g/mol) 244.33
Synonym Bis-ε-aminocaproic Acid, N-(6'-Aminocaproyl)-6-aminocaproic Acid, 6-Aminohexanoic Acid Dimer, 6-[(6-Amino-1-oxohexyl)amino]hexanoic Acid;
IUPAC Name 6-(6-aminohexanoylamino)hexanoic acid
SMILES OC(CCCCCNC(CCCCCN)=O)=O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C12H24N2O3/c13-9-5-1-3-7-11(15)14-10-6-2-4-8-12(16)17/h1-10,13H2,(H,14,15)(H,16,17)
Formula Weight 244.1787
Percent Purity >95%
Chemical Name or Material 6-Aminocaproic Acid Dimer
Grade HPLC