Learn More
6-Acetamidohexanoic acid hydrate, 99%
Product Code. 11475657
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11475657
Brand: Thermo Scientific Alfa Aesar B23623.14
Request Bulk or Custom Format
CAS: 57-08-9 | C8H15NO3 | 173.21 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 57-08-9
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00004424
InChI Key WDSCBUNMANHPFH-UHFFFAOYSA-N
Synonym acexamic acid, 6-acetylamino hexanoic acid, 6-acetamidocaproic acid, plastenan, acetaminocaproic acid, acide acexamique, aeah base, hexanoic acid, 6-acetylamino, acemin, epsilon-acetamidocaproic acid
PubChem CID 2005
IUPAC Name 6-acetamidohexanoic acid
SMILES CC(=O)NCCCCCC(O)=O
Specifications
Melting Point 102°C to 106°C
Linear Formula CH3CONH(CH2)5CO2H·xH2O
Quantity 25 g
Merck Index 14,45
Formula Weight 173.21 (Anhydrous)
Percent Purity 99%
Chemical Name or Material 6-Acetamidohexanoic acid hydrate