missing translation for 'onlineSavingsMsg'
Learn More

6,8-Dimethoxy Moxifloxacin Hydrochloride, TRC

Product Code. 30376012
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30376012 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376012 Supplier Toronto Research Chemicals Supplier No. TRCD47060010MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6,8-Dimethoxy Moxifloxacin hydrochloride
Molecular Formula C22 H27 N3 O5 . x H Cl
Quantity 10 mg
Synonym 1-Cyclopropyl-1,4-dihydro-6,8-dimethoxy-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic Acid Hydrochloride, Moxifloxacin EP Impurity B HCl Salt
InChI Formula InChI=1S/C22H27N3O5/c1-29-17-8-14-18(25(13-5-6-13)10-15(20(14)26)22(27)28)21(30-2)19(17)24-9-12-4-3-7-23-16(12)11-24/h8,10,12-13,16,23H,3-7,9,11H2,1-2H3,(H,27,28)
SMILES COc1cc2C(=O)C(=CN(C3CC3)c2c(OC)c1N4CC5CCCNC5C4)C(=O)O
IUPAC Name 7-(1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl)-1-cyclopropyl-6,8-dimethoxy-4-oxo-quinoline-3-carboxylic acid
Molecular Weight (g/mol) 413.47
Formula Weight 413.1951
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.