missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydrocanrenone, TRC

Product Code. 18042730
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18042730 100 mg 100mg
17929849 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18042730 Supplier Toronto Research Chemicals Supplier No. TRCD448720100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 976-70-5
Molecular Formula C22 H30 O3
Molecular Weight (g/mol) 342.47
Synonym Pregn-4-ene-21-carboxylic acid, 17-hydroxy-3-oxo-, γ-lactone, (17α)-, 17α-Pregn-4-ene-21-carboxylic acid, 17-hydroxy-3-oxo-, γ-lactone (6CI,7CI,8CI), 17β-Hydroxy-3-oxopregn-4-en-21-carboxylic acid γ-lactone, 3-(3-Oxo-17β-hydroxy-4-androsten-17-yl)propionic acid γ-lactone, 3-(3-Oxo-17β-hydroxyandrost-4-en-17α-yl)propiolactone, 3'-(3-Oxo-17β-hydroxyandrost-4-en-17α-yl)-propionic acid lactone, 6,7-Dihydrocanrenone, SC 5233, (2′R)-3′,4′-Dihydro-5′H-spiro[androst-4-ene-17,2′-furan]-3,5′-dione, Aldone
IUPAC Name (8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione
SMILES C[C@]12CCC(=O)C=C1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@@]45CCC(=O)O5

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H30O3/c1-20-9-5-15(23)13-14(20)3-4-16-17(20)6-10-21(2)18(16)7-11-22(21)12-8-19(24)25-22/h13,16-18H,3-12H2,1-2H3/t16-,17+,18+,20+,21+,22-/m1/s1
Formula Weight 342.2195
Chemical Name or Material 6,7-Dihydrocanrenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.