missing translation for 'onlineSavingsMsg'
Learn More

6,6'-Dibromo-3,3'-bipyridine, tech. 90%, Thermo Scientific™

Product Code. 11332859
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11332859

Brand: Thermo Scientific Alfa Aesar H27726.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147496-14-8
Molecular Formula C10H6Br2N2
Molecular Weight (g/mol) 313.98
MDL Number MFCD09037441
InChI Key IJTQXCQPBVBRSV-UHFFFAOYSA-N
Synonym 6,6'-dibromo-3,3'bipyridine, 6,6'-dibromo-3,3'-bipyridine, 2-bromo-5-6-bromopyridin-3-yl pyridine, 6,6'-dubromo 3,3' bipyridil, acmc-20am0r, 6,6'-dibromo-3,3' bipyridine, 3,3'-bipyridine,6,6'-dibromo, 6,6-dibromo-3,3-bipyridine,tech, 6,6'-dibromo-3,3'-bipyridine, tech
PubChem CID 6426300
SMILES BrC1=NC=C(C=C1)C1=CN=C(Br)C=C1

Specifications

Melting Point ∽249°C (decomposition)
Quantity 100 mg
IUPAC Name 2-bromo-5-(6-bromopyridin-3-yl)pyridine
Formula Weight 313.98
Percent Purity 90%
Grade Technical
Chemical Name or Material 6,6'-Dibromo-3,3'-bipyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.