Learn More
6-[2-Biotinylamidoethyl]-dithiopropionamido]-4,8-diaza-5,7-diketoundecanoic Acid Bis-N-sulfosuccinimidyl Ester Disodium Salt, TRC
Product Code. 30342522
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30342522 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30342522 Supplier Toronto Research Chemicals Supplier No. TRCB3927002MG
 Restricted Item
Estimated Shipment: 09-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-[2-Biotinylamidoethyl]-dithiopropionamido]-4,8-diaza-5,7-d
CAS 1044220-56-5
Molecular Formula C32 H42 N8 O19 S5 . 2 Na
Quantity 2 mg
Synonym 6-[2-Biotinylamidoethyl]-Dithiopropionamido]-4,8-Diaza-5,7-Diketoundecanoic Acid, Bis-N-Sulfosuccinimidyl Ester Disodium Salt
InChI Formula InChI=1S/C32H44N8O19S5.2Na/c41-20(4-2-1-3-17-26-16(15-60-17)36-32(51)38-26)33-10-12-62-61-11-7-21(42)37-27(28(47)34-8-5-24(45)58-39-22(43)13-18(30(39)49)63(52,53)54)29(48)35-9-6-25(46)59-40-23(44)14-19(31(40)50)64(55,56)57;;/h16-19,26-27H,1-15H2,(H,33,41)(H,34,47)(H,35,48)(H,37,42)(H2,36,38,51)(H,52,53,54)(H,55,56,57);;/q;2*+1/p-2/t16-,17-,18?,19?,26-,27?;;/m0../s1
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1CC(=O)N(OC(=O)CCNC(=O)C(NC(=O)CCSSCCNC(=O)CCCC[C@@H]2SC[C@@H]3NC(=O)N[C@H]23)C(=O)NCCC(=O)ON4C(=O)CC(C4=O)S(=O)(=O)[O-])C1=O
IUPAC Name disodium;1-[3-[[2-[3-[2-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]ethyldisulfanyl]propanoylamino]-3-[[3-(2,5-dioxo-3-sulfonatopyrrolidin-1-yl)oxy-3-oxopropyl]amino]-3-oxopropanoyl]amino]propanoyloxy]-2,5-dioxopyrrolidine-3-sulfonate
Molecular Weight (g/mol) 1049.02
Formula Weight 1048.0965