missing translation for 'onlineSavingsMsg'
Learn More

6-(2,2,2-Trifluoroethoxy)pyrimidine-4-carboxylic Acid, TRC

Product Code. 30415408
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415408 1 g 1g
30427862 100 mg 100mg
30423944 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30415408 Supplier Toronto Research Chemicals Supplier No. TRCT2693911G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1439896-74-8
Molecular Formula C7H5F3N2O3
Molecular Weight (g/mol) 222.12
IUPAC Name 6-(2,2,2-trifluoroethoxy)pyrimidine-4-carboxylic acid
SMILES OC(=O)c1cc(OCC(F)(F)F)ncn1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H5F3N2O3/c8-7(9,10)2-15-5-1-4(6(13)14)11-3-12-5/h1,3H,2H2,(H,13,14)
Formula Weight 222.0252
Chemical Name or Material 6-(2,2,2-Trifluoroethoxy)pyrimidine-4-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.