Learn More
(5S)-N-Benzyloxycarbonyl-N-(5-Amino-1-carboxypentyl)iminodiacetic Acid, Tri-t-butyl Ester, TRC
Product Code. 30336153
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336153 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30336153 Supplier Toronto Research Chemicals Supplier No. TRCB285896250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Benzyloxycarbonyl-N-(5-Amino-1-carboxypentyl)iminodiacetic
CAS 205379-07-3
Molecular Formula C30 H48 N2 O8
Quantity 250 mg
InChI Formula InChI=1S/C30H48N2O8/c1-28(2,3)38-24(33)19-32(20-25(34)39-29(4,5)6)23(26(35)40-30(7,8)9)17-13-14-18-31-27(36)37-21-22-15-11-10-12-16-22/h10-12,15-16,23H,13-14,17-21H2,1-9H3,(H,31,36)/t23-/m0/s1
SMILES CC(C)(C)OC(=O)CN(CC(=O)OC(C)(C)C)[C@@H](CCCCNC(=O)OCc1ccccc1)C(=O)OC(C)(C)C
IUPAC Name tert-butyl (2S)-2-[bis[2-[(2-methylpropan-2-yl)oxy]-2-oxoethyl]amino]-6-(phenylmethoxycarbonylamino)hexanoate
Molecular Weight (g/mol) 564.711
Formula Weight 564.341