Learn More
(5R)-Tetrahydro-5-methyl-2H-pyran-2-one, TRC
Product Code. 30428617
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428617 5 g 5g
30417184 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30428617 Supplier Toronto Research Chemicals Supplier No. TRCT2939705G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2857-75-2
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
Synonym (R)-Tetrahydro-5-methyl-2H-pyran-2-one
IUPAC Name (5R)-5-methyloxan-2-one
SMILES C[C@@H]1CCC(=O)OC1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C6H10O2/c1-5-2-3-6(7)8-4-5/h5H,2-4H2,1H3/t5-/m1/s1
Formula Weight 114.0681
Chemical Name or Material (5R)-Tetrahydro-5-methyl-2H-pyran-2-one