missing translation for 'onlineSavingsMsg'
Learn More

Xenazoic Acid, TRC

Product Code. 30453220
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30453220

Brand: Toronto Research Chemicals TRCX7444505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Xenazoic Acid
CAS 1174-11-4
Molecular Formula C23H21NO4
Quantity 5 mg
Synonym p-[(alpha-Ethoxy-p-Phenylphenacyl)Amino]Benzoic Acid, CV 58903, NSC 43842, NSC 58991, NSC 59182, SKF 8318, Xanalamine, Xenalamine, Xenovis, 4-[(2-[1,1'-Biphenyl]-4-yl-1-Ethoxy-2-Oxoethyl)Amino]-Benzoic acid
InChI Formula InChI=1S/C23H21NO4/c1-2-28-22(24-20-14-12-19(13-15-20)23(26)27)21(25)18-10-8-17(9-11-18)16-6-4-3-5-7-16/h3-15,22,24H,2H2,1H3,(H,26,27)
SMILES CCOC(Nc1ccc(cc1)C(=O)O)C(=O)c2ccc(cc2)c3ccccc3
IUPAC Name 4-[[1-ethoxy-2-oxo-2-(4-phenylphenyl)ethyl]amino]benzoic acid
Molecular Weight (g/mol) 375.42
Formula Weight 375.1471
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.