missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Velneprit, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Velneprit |
| CAS | 342577-38-2 |
| Molecular Formula | C17H24F3N3O3S |
| Quantity | 5 mg |
| Synonym | N-[5-(Trifluoromethyl)pyridin-2-yl]-trans-4-(tert-butylsulfonylamino)cyclohexane-1-carboxamide, S 2367 |
| InChI Formula | InChI=1S/C17H24F3N3O3S/c1-16(2,3)27(25,26)23-13-7-4-11(5-8-13)15(24)22-14-9-6-12(10-21-14)17(18,19)20/h6,9-11,13,23H,4-5,7-8H2,1-3H3,(H,21,22,24) |
| SMILES | CC(C)(S(=O)(N[C@H]1CC[C@H](C(NC2=NC=C(C(F)(F)F)C=C2)=O)CC1)=O)C |
| Molecular Weight (g/mol) | 407.45 |
| Formula Weight | 407.149 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction