Learn More
S-(-)-Warfarin, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | (S)-Warfarin |
| CAS | 5543-57-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 H16 O4 |
| Quantity | 5 mg |
| Synonym | 2H-1-Benzopyran-2-one','4-hydroxy-3-[(1S)-3-oxo-1-phenylbutyl]-, 2H-1-Benzopyran-2-one','4-hydroxy-3-(3-oxo-1-phenylbutyl)-','(S)-, Coumarin','3-(α-acetonylbenzyl)-4-hydroxy-','(S)-(-)- (8CI), 4-Hydroxy-3-[(1S)-3-oxo-1-phenylbutyl]-2H-1-benzopyran-2-one, (-)-Warfarin, (S)-Warfarin |
| InChI Formula | InChI=1S/C19H16O4/c1-12(20)11-15(13-7-3-2-4-8-13)17-18(21)14-9-5-6-10-16(14)23-19(17)22/h2-10,15,21H,11H2,1H3/t15-/m0/s1 |
| SMILES | CC(=O)C[C@H](C1=C(O)c2ccccc2OC1=O)c3ccccc3 |
| IUPAC Name | 4-hydroxy-3-[(1S)-3-oxo-1-phenylbutyl]chromen-2-one |
| Molecular Weight (g/mol) | 308.33 |
| Show More |