missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Janelia Fluor™ 646, free acid

Product Code. 30484803
Click to view available options
:
5 mg
This item is not returnable. View return policy

Product Code. 30484803

Brand: Tocris Bioscience™ 8915/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Red dye (Reactive group: free acid). Application: flow cytometry, confocal microscopy, super resolution microscopy (SRM) including dSTORM & STED. Suitable for live cell imaging

Key Information: Janelia Fluor™ 646, free acid is a red fluorogenic fluorescent dye; supplied with a free acid reactive group. Suitable for live cell imaging.Application: Suitable for flow cytometry, confocal microscopy, super resolution microscopy (SRM) including dSTORM (in both live and fixed cells) and STED. Used in protocol (2017 Grimm et al - see references below) for the synthesis of Janelia Fluor™ HaloTag™ and SNAP-Tag™ ligands, for use in live cell imaging experiments. Cell permeable.Properties and Photophysical Data: Excitation and emission maxima (λ) are 646 nm and 664 nm, respectively; quantum yield = 0.54; extinction coefficient = 152,000 M-1cm-1. A280 correction factor = 0.19; lactone-zwitterion equilibrium constant (KL-Z) = 0.0012. To measure the absorbance spectrum of this dye we recommend the following solvent: ethanol or trifluoroethanol plus 0.1% TFA. We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company Novus Biologicals. HaloTag is a trademark of Promega Corporation, and SNAP-tag is a trademark of New England BioLabs, Inc. Janelia Fluor is a registered trademark of Howard Hughes Medical Institute. This product will replace Janelia Fluor™ 646, free acid (Cat. No. 6993).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[7-(1-Azetidinyl)-10-(2,5-dicarboxyphenyl)-9,9-dimethyl-9-silaanthracen-2(9H)-ylidene]azetidinium inner salt
CAS 1811539-57-7
Quantity 5 mg
Target Free Acid Janelia Fluor Dyes
Molecular Formula C29H28N2O4Si
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.