Learn More
5alpha-Androstan-3beta,17alpha-diol, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | 5alpha-Androstan-3beta,17alpha-diol |
| CAS | 5856-11-1 |
| Molecular Formula | C19 H32 O2 |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C19H32O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12-17,20-21H,3-11H2,1-2H3/t12-,13-,14-,15-,16-,17+,18-,19-/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CC[C@H]4C[C@@H](O)CC[C@]34C)[C@@H]1CC[C@H]2O |
| IUPAC Name | (3S,5S,8R,9S,10S,13S,14S,17R)-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene-3,17-diol |
| Molecular Weight (g/mol) | 292.46 |