missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,14,14-Tetramethyl-8-oxopentadecanedioic Acid-d4, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | 2,2,14,14-Tetramethyl-8-oxopentadecanedioic Acid-d4 |
| CAS | 2408132-01-2 |
| Molecular Formula | C19 D4 H30 O5 |
| Quantity | 5 mg |
| Synonym | 8-Oxo-2,2,14,14-tetramethylpentadecanedioic Acid-d4, ESP 15228-d4 |
| InChI Formula | InChI=1S/C19H34O5/c1-18(2,16(21)22)13-9-5-7-11-15(20)12-8-6-10-14-19(3,4)17(23)24/h5-14H2,1-4H3,(H,21,22)(H,23,24)/i11D2,12D2 |
| SMILES | [2H]C([2H])(CCCCC(C)(C)C(=O)O)C(=O)C([2H])([2H])CCCCC(C)(C)C(=O)O |
| IUPAC Name | 7,7,9,9-tetradeuterio-2,2,14,14-tetramethyl-8-oxopentadecanedioic acid |
| Molecular Weight (g/mol) | 346.495 |
| Formula Weight | 346.266 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction