missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1,2-ethanediyl)bis-Carbamodithioic Acid Dimethyl Ester, TRC

Product Code. 17953715
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17953715 5 mg 5mg
30134408 10 mg 10mg
17921005 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17953715 Supplier Toronto Research Chemicals Supplier No. TRCM6212005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80555-60-8
Molecular Formula C7H14N2S4
Molecular Weight (g/mol) 254.46
Synonym S,S'-Dimethylxanthogenpropylenebisdithiocarbamate, Dimethylxanthogenpropylenebisdithiocarbamate: S,S'-Dimethylxanthogen-1,2-propylenebisdithiocarbamate
IUPAC Name methyl N-[1-(methylsulfanylcarbothioylamino)propan-2-yl]carbamodithioate
SMILES CSC(=S)NCC(C)NC(=S)SC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C7H14N2S4/c1-5(9-7(11)13-3)4-8-6(10)12-2/h5H,4H2,1-3H3,(H,8,10)(H,9,11)
Formula Weight 254.004
Percent Purity >95%
Chemical Name or Material (1-Methyl-1,2-ethanediyl)bis-Carbamodithioic Acid Dimethyl Ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.