Learn More
Chem-Lab 5LT Potassium iodide-iodate 0.2 N sol. (33.2g KI+ 7.134g KIO3 + 0.24g NaOH / l H2O (±0.
Brand: Chem-Lab CL05.1140.5000
5LT Potassium iodide-iodate 0.2 N sol. (33.2g KI+ 7.134g KIO3 + 0.24g NaOH / l H2O (±0.