Learn More
JT Baker 5KG AMMONIUM PEROXODISULFATE BAKER ANALYZED Reagent
Brand:  JT Baker 0762.5000
5KG AMMONIUM PEROXODISULFATE BAKER ANALYZED Reagen
Product Code. 15517864
  • ONLINE EXCLUSIVE PRICE
    309.00€
    219.59€ / 5kg
    Save 89.41€
    28% Off
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.