missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benomyl, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Benomyl |
| CAS | 17804-35-2 |
| Molecular Formula | C14 H18 N4 O3 |
| Quantity | 5 g |
| Synonym | 2-Benzimidazolecarbamic acid, 1-(butylcarbamoyl)-, methyl ester (8CI), Carbamic acid, [1-[(butylamino)carbonyl]-1H-benzimidazol-2-yl]-, methyl ester (9CI), Agrocit, BBC, BC 6597, Benfungin, Benfungin WP, Benlate, Benlate 50, Benlate 50W, Benlate 50WP, Benlate WP, Benomil, Benomyl, Benomyl-Imex, Du Pont 1991, Fundazol, Fundazol 50WP, Fungicide D-1991, Fungochrom, Kribenomyl, MBC, Marvel, Methyl 1-(butylcarbamoyl)-2-benzimidazolecarbamate, Methyl 1-(butylcarbamoyl)-2-benzimidazolylcarbamate, NS 02, NS 02 (fungicide), NSC 263489, Safomyl, Tersan 1991, Uzgen, Zetamil |
| InChI Formula | InChI=1S/C14H18N4O3/c1-3-4-9-15-13(19)18-11-8-6-5-7-10(11)16-12(18)17-14(20)21-2/h5-8H,3-4,9H2,1-2H3,(H,15,19)(H,16,17,20) |
| SMILES | CCCCNC(=O)n1c(NC(=O)OC)nc2ccccc12 |
| IUPAC Name | methyl N-[1-(butylcarbamoyl)benzimidazol-2-yl]carbamate |
| Molecular Weight (g/mol) | 290.32 |
| Formula Weight | 290.1379 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction