Learn More
Aniline-2,3,4,5,6-d5, CDN
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Aniline D5 |
| CAS | 4165-61-1 |
| Molecular Formula | C6 2H5 H2 N |
| Quantity | 5 g |
| InChI Formula | InChI=1 S/C6H7N/c7-6-4-2-1-3-5-6/h1-5 H,7H2/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(N)c([2 H])c1[2 H] |
| IUPAC Name | 2,3,4,5,6-pentadeuterioaniline |
| Molecular Weight (g/mol) | 98.16 |
| Formula Weight | 98.0892 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |