missing translation for 'onlineSavingsMsg'
Learn More

5Beta-Pregnan-3Alpha-ol-20-one, TRC

Product Code. 30421632
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30421632 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30421632 Supplier Toronto Research Chemicals Supplier No. TRCP7113005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5B-Pregnan-3a-ol-20-one
CAS 128-20-1
Purity Grade Notes HPLC
Molecular Formula C21H34O2
Quantity 5 mg
Synonym Pregnan-20-one','3-hydroxy-','(3α,5β)-, 5β-Pregnan-20-one','3α-hydroxy- (6CI,8CI), (3α,5β)-3-Hydroxypregnan-20-one, 3-Deoxo-3α-hydroxy-5β-dihydroprogesterone, 3α,5β-Pregnanolone, 3α,5β-Tetrahydroprogesterone, 3α-Hydroxy-20-oxo-5β-pregnane, 3α-Hydroxy-5β,17β-pregnan-20-one, 3α-Hydroxy-5β-pregnan-20-one, 3α-Hydroxy-5β-tetrahydroprogesterone, 5β-Pregnan-3α-ol-20-one, 5β-Pregnane-3α-ol-20-one, Eltanolone, KABI 2213, NSC 82867, Pregnan-3α-ol-20-one, Pregnanolone, Pregnanolone II, SKF 6455, U 2340
InChI Formula InChI=1S/C21H34O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h14-19,23H,4-12H2,1-3H3/t14-,15-,16+,17-,18+,19+,20+,21-/m1/s1
SMILES CC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name 1-[(3R,5R,8R,9S,10S,13S,14S,17S)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
Molecular Weight (g/mol) 318.5
Formula Weight 318.2559
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.