Learn More
5alpha-Pregnan-17alpha,21-diol-3,11,20-trione-1,2,4,5-d4, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 5 alpha-Dihydrocortisone-1,2,4,5-D4 |
| Molecular Formula | C21 D4 H26 O5 |
| Quantity | 0.005 g |
| Synonym | 17,21-Dihydroxy-5α-pregnane-3,11,20-trione-1,2,4,5-d4, 17α,21-Dihydroxy-5α-pregnane-3,11,20-trione-1,2,4,5-d4, 5α-Dihydrocortisone-1,2,4,5-D4, 5α-Pregnane-17α,21-diol-3,11,20-trione-1,2,4,5-d4, Allodihydrocortisone-1,2,4,5-d4 |
| InChI Formula | InChI=1 S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h12,14-15,18,22,26 H,3-11H2,1-2H3/t12-,14-,15-,18+,19-,20-,21-/m0/s1/i5D,7 D,9 D,12 D/t5?,7?,9?,12-,14-,15-,18+,19-,20-,21- |
| SMILES | [2 H]C1C([2 H])[C@]2(C)[C@@H]3 [C@@H](CC[C@@]2([2 H])C([2 H])C1=O)[C@@H]4 CC[C@](O)(C(=O)CO)[C@@]4(C)CC3=O |
| IUPAC Name | (5 S,8 S,9 S,10 S,13 S,14 S,17 R)-1,2,4,5-tetradeuterio-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,4,6,7,8,9,12,14,15,16-decahydro-1 H-cyclopenta[a]phenanthrene-3,11-dione |
| Molecular Weight (g/mol) | 366.4846 |
| Formula Weight | 366.2344 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |